Friedman Bone Rongeur Forceps, Sklar
Supplier: Sklar
40-3190 40-3192
82030-198EA 747.07 CAD
82030-198 82030-200
Friedman Bone Rongeur Forceps, Sklar
Bone Cutters and Rongeurs
Friedman Bone Rongeur is a double spring, plier handle instrument used for debulking and removing pieces of bone
This rongeur has delicate jaws for use with delicate bones such as ear, nasal, and orbital. The slight curvature of the jaws makes this instrument useful in these obscure areas as well.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
SPECIFICATIONS
Disposable / reusable Reusable
Environmentally Preferable
Latex/Latex-free Latex-free