Tru-Punch™ Sterile Disposable Biopsy Punches, Sklar
Supplier: Sklar
96-1098 96-1100 96-1105 96-1110 96-1115 96-1120 96-1125 96-1130 96-0102 96-0192 96-1097 96-1102 96-1144 96-1160 96-1150 96-1148 96-1152 96-1146 96-1140 96-1142 96-0160 96-1092
82030-340PK 416.01 CAD
82030-340 82030-342 82030-344 82030-346 82030-348 82030-350 82030-352 82030-354 10185-236 10185-238 10185-260 10185-262 10185-264 10192-324 10192-382 10192-404 10192-424 10193-710 10193-796 10193-846 10193-970 75995-668
Tru-Punch™ Sterile Disposable Biopsy Punches, Sklar
Biopsy Punches
Single-use dermal biopsy punches.
  • Seamless steel tip honed for a razor-sharp incision
  • Hollow, ribbed polypropylene handle for easy removal of tissue sample
Assorted Box of 25 contains 5 each of 2mm, 3mm, 3.5mm, 4mm and 5mm. Assorted Box of 50 contains 10 each of 2mm, 3mm, 3.5mm, 4mm and 5mm.
Order Now
SPECIFICATIONS
Environmentally Preferable