SP Bel-Art Sterileware® Long Handle Sterile Polystyrene Sampling Spoons
Supplier: Bel-Art Products, a Part of SP
H36940-0000 H36942-0000 369440000 H36944-0010 H36946-0000 H36940-0010 H36942-0010 H36946-0010
56925-109PK 567.47 CAD
56925-109 56925-131 56925-164 89140-582 56925-197 89140-578 89140-580 89140-584
SP Bel-Art Sterileware® Long Handle Sterile Polystyrene Sampling Spoons
Scoops
Designed to gather small amounts of sample material, these sterile, polystyrene plastic sampling spoons are available in four bowl sizes.
  • Long-handled design
  • Sterilized by gamma irradiation
  • Polystyrene plastic construction
  • Lot stamped for accurate lot tracing
The long handle reaches easily into jars, bottles, and other confined areas, and minimizes the potential of personal contact with sampled material.
All spoons are FDA-grade and safe for food, drug, and cosmetic contact. All sterile products are individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.
Packaging: Please note that the sterile packaging bag was previously 9L×3"W, and the new dimensions are 12.5L×5.38"W.