Nalgene® Square Media Bottles, PETG, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
342020-0030 342020-0125 342020-0500 342020-1000 342020-0250 342020-0060 342020-2000 342020-9500 342024-0030 342024-0060 342024-0125 342024-0250 342024-0500 342024-1000 342020-0060 342020-2000
16059-490CS 1323.54 CAD
16059-490 16059-494 16059-496 16059-498 16059-912 16059-914 16159-999 21010-402 16057-302 16057-304 16057-306 16057-308 16057-310 16057-314 CATX16059-914 CATX16159-999
Nalgene® Square Media Bottles, PETG, Sterile, Thermo Scientific
Bottles Plastic Bottles
Nalgene® polyethylene terephthalate copolyester (PETG) square media bottles are an inexpensive alternative to glass media bottles.
  • Available with or without a natural HDPE screw cap closure
  • Bottles and closures are not autoclavable
  • Break-resistant, durable, and leakproof
Transparency and reduced permeability to CO₂/O₂ allow media storage for up to six months. Bottles and closures are radiation-sterilized and nonpyrogenic. Bottle 16159-999 (2 L size) features molded-in handgrips.
Ordering information: Packaged in shrink-wrapped trays. For additionalMore Product Information