VWR® Premium Aluminum Foil
Supplier: VWR International
89107-724 89107-726 89107-730 89107-732 89107-734 89107-736
89107-724CS 222.07 CAD
89107-724 89107-726 89107-730 89107-732 89107-734 89107-736
VWR® Premium Aluminum Foil
Aluminium Foils
Premium, ultra-clean aluminum foil is strong and puncture-resistant.
  • Available in a variety of formats
  • Contains 200 sheets
  • Puncture-resistant
Foil is able to withstand extreme heat and cold, and has the lowest moisture-vapor transfer rate of all wrapping materials. Standard, heavy-duty, and extra heavy-duty formats are supplied as boxed rolls with a metal tear-off bar for ease in obtaining exact size requirements. The pop-up dispenser contains 200 individual sheets.
Order Now
SPECIFICATIONS
Environmentally Preferable