LabFit™ Base Cabinet Doors
Supplier: TrippNT
53672 53677 53659 53630 53667 53669 53670 53671 53682 53678 53679 53680 53681
77135-322EA 3953.02 CAD
77135-322 77135-332 77135-298 77135-266 77135-312 77135-316 77135-318 77135-320 77135-342 77135-334 77135-336 77135-338 77135-340
LabFit™ Base Cabinet Doors
Furniture Casework
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
  • Non-metal construction
  • Resistant to chemicals and corrosion
  • Easy to clean and disinfect
  • Large removeable rear access panel
  • Embedded magnetic door latches
  • Industry leading two year full replacement warranty
  • Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistnace to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it an ideal option for a range of laboratory and medical applications. Joints are welded insuring a leak-tight, flush union. All hinges, handles, drawer slides, and stops are constructed of polypropylene.
Delivery information: TrippNT® LabFit™ laboratory casework has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor furniture specialist.