LabFit™ Base Cabinet Drawers
Supplier: TrippNT
53673 53674 53651 53652 53653 53660 53648 53683 53684 53685
77135-324EA 5124.95 CAD
77135-324 77135-326 77135-284 77135-286 77135-288 77135-300 77135-278 77135-344 77135-346 77135-348
LabFit™ Base Cabinet Drawers
Furniture Casework
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
  • Non-metal construction
  • Resistant to chemicals and corrosion
  • Easy to clean and disinfect
  • Large removeable rear access panel
  • Industry leading two year full replacement warranty
  • Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistance to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it an ideal option for a range of laboratory and medical applications. Joints are welded insuring a leak-tight, flush union. All hinges, handles, drawer slides, and stops are constructed of polypropylene.
Delivery information: TrippNT® LabFit laboratory casework has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor furniture specialist.