Stilla® naica® Systems, 3-color
Supplier: Stilla Technologies
Stilla®
N10002.3 N10001.3
77392-272EA 76845.83 CAD
77392-272 77392-274
Stilla® naica® Systems, 3-color
Thermal Cyclers
The 3-color naica® system is an easy-to-use digital PCR platform that supports 3 colors and harnesses cutting-edge microfluidic technology to integrate the digital PCR workflow onto a single consumable chip.
  • Easy-to-use digital PCR solution, with minimal hands-on time
  • Up to 3-color multiplexing, from 12 to 48 samples/run
  • Fast time to results
The 3-color naica® system integrates the digital PCR process in a single consumable, reducing hands-on time and interactions, to simply insert your sample and walk away. The 3-color naica® system utilizes Crystal Digital PCR® technology, which partitions samples into a large array of thousands of individual droplets - each its own reaction compartment - before amplifying nucleic acid molecules in each droplet, for the absolute quantification of nucleic acids based on cutting-edge microfluidic technology. These reactions are tagged with fluorophores to be read using up to three different fluorescence light channels.