SingleQuots® MEGM® Mammary Epithelial Cell Growth Medium Kit
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-4136
CA10063-936EA 223.18 CAD
CA10063-936
SingleQuots® MEGM® Mammary Epithelial Cell Growth Medium Kit
Cell Culture Media
MEGM® SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the MEGM® Mammary Epithelial Cell Growth Medium BulletKit® (Catalog Number CC-3150).
The full MEGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3150.
Mammary Epithelial Cell Growth Medium BulletKit® has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research.
Delivery information: Content:
1 × Orange Cap Vial with BPE 2.00 ml
1 × Green Cap Vial with hEGF 0.50 ml
1 × Lilac Cap Vial with Insulin 0.50 ml
1 × Natural Cap Vial with Hydrocortisone 0.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml