Bovine Pituitary Extract (BPE)
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-4009
CA10063-922EA 211.33 CAD
CA10063-922
Bovine Pituitary Extract (BPE)
Cell Culture Supplements and Additives
Bovine Pituitary Extract is useful in the culture of various epithelial cell types. The product is included in SingleQuots® Kits associated with various media systems for epithelial cell types. The product is available at a concentration of 13 mg/ml.
Delivery information: Content: 1 × Vial Containing 13 mg/ml BPE 2 ml.