SKBM®- 2 Skeletal Muscle Cell Growth Basal Medium-2
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3246
CA10063-908EA 258.73 CAD
CA10063-908
SKBM®- 2 Skeletal Muscle Cell Growth Basal Medium-2
Cell Culture Media
SkBM® -2 is a Skeletal Muscle Myoblast basal medium sold separately. This can be used with 1 kit of CC-3244 (skeletal muscle myoblast growth medium SingleQuots® Supplements and Growth Factors).
The full SkGM® -2 BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3245.
SkGM®-2 Skeletal Muscle Cell Growth Medium-2 (catalog number CC-3245) is an enhancement of SkGM® Medium. Fetuin has been removed and there has been further optimization of the basal media.
SkGM® Medium is optimized for use with human skeletal muscle cells. SkGM® -2 is specially designed for use with normal and diabetic skeletal muscle myoblasts.
Delivery information: Content: 1 × Bottle Basal Medium 500 ml