SingleQuots® SKGM® - 2 Skeletal Muscle Cell Growth Medium-2 Supplements and Growth Factors
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3244
CA10063-904EA 395 CAD
CA10063-904
SingleQuots® SKGM® - 2 Skeletal Muscle Cell Growth Medium-2 Supplements and Growth Factors
Cell Culture Media
SkGM® -2 SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the SkGM® -2 Skeletal Muscle Myoblast Medium BulletKit® (Catalog Number CC-3245).
The full SkGM® -2 BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3245.
SkGM®-2 Skeletal Muscle Cell Growth Medium-2 is an enhancement of SkGM® Medium. Fetuin has been removed and there has been further optimization of the basal media.
SkGM® Medium is optimized for use with human skeletal muscle cells. SkGM® -2 is specially designed for use with normal and diabetic skeletal muscle myoblasts.
Delivery information: Content:
1 × Red Cap Vial with GA-1000 0.50 ml
1 × Green Cap Vial with hEGF 0.50 ml
1 × Natural Cap Vial with Dexamethasone 0.50 ml
1 × Bottle with FBS 50.00 ml
1 × Bottle L-Glutamine 10.00 ml