BulletKit® CDM® Chondrocyte Differentiation Medium
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3225
CA10063-890EA 1042.8 CAD
CA10063-890
BulletKit® CDM® Chondrocyte Differentiation Medium
Cell Culture Media
CDM® Chondrocyte Differentiation Medium is specially designed for the differentiation of chondrocytes. The BulletKit® is conveniently packaged with basal medium and supplements included for the differentiation of chondrocytes.
CGM® Chondrocyte Growth Medium is also available and sold separately for expansion of human chondrocytes.
Delivery information: Content:
1 × Bottle Basal Medium 250 ml
1 × Yellow Cap Vial with R3-IGF 0.50 ml
1 × Blue Cap Vial with TGF-beta 1.25 ml
1 × Lilac Cap Vial with Insulin 0.50 ml
1 × Red Cap Vial with GA-1000 0.25 ml
1 × Bottle FBS 12.50 ml
1 × Natural Cap Vial with Transferrin 0.50 ml
Order Now
SPECIFICATIONS
Environmentally Preferable