NPBM® Neural Progenitor Basal Medium
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3210
CA10064-354EA 221.2 CAD
CA10064-354
NPBM® Neural Progenitor Basal Medium
Cell Culture Media
Poietics® Neural Progenitor Cell System contains Norman Human Neural Progenitor Cells (NHNP) and defined medium optimized for their growth and in vitro differentiation.
This system may be used for experimental applications including drug development, neurotoxicity, neurogenesis, electrophysiology, neurotransmitter disorders, and central nervous system function.
The available defined neural progenitor media includes a growth medium BulletKit® (CC-3209) as well as a differentiaion medium BulletKit® (CC-3229).
Poietics® Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.
Delivery information: Content: Neural Progenitor Basal Medium (1 bottle).