MEBM® Mammary Epithelial Cell Growth Basal Medium
Supplier: Lonza
CC-3151
CA10065-260EA 242.93 CAD
CA10065-260
MEBM® Mammary Epithelial Cell Growth Basal Medium
Cell Culture Media
MEBM® is a mammary epithelial cell basal medium sold separately. This can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots® Supplements and Growth Factors). The full MEGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3150.
MEGM® Mammary Epithelial Cell Growth Medium BulleKit® has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research.
Delivery information: Content: 1 × Bottle Basal Medium 500 ml.