BulletKit® MEGM® Mammary Epithelial Cell Growth Medium
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3150
CA101968-208KT 381.18 CAD
CA101968-208
BulletKit® MEGM® Mammary Epithelial Cell Growth Medium
Cell Culture Media
MEGM® Mammary Epithelial Cell Growth Medium has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research.
MEGM® BulletKit® is optimized and readily available media kit giving researchers the confidence and the convenience of a system that works to support their cells. This is especially important for researchers who are utilizing normal breast cell lines as a stepping stone to primary HMEC cell culture. Working with the same media between cell lines and primary cells can reduce variability within the experiments and simplifies the shift for researchers. In addition, if primary HMECs are utilized as a side-by-side control with normal breast cell lines, the MEGM® BulletKit® media may also offer cost-benefits since customers can use the same media to support both cell types.
MEGM®Bulletkit®, is also sold as separate basal medium and supplements.
Delivery information: Content:
1 × MEBM® Basal Medium (CC-3151), 500 ml
1 × MEGM® SingleQuots® Supplement Pack (CC-4136) containing:
1 × Orange Cap Vial with BPE, 2.00 ml
1 × Green Cap Vial with hEGF, 0.50 ml
1 × Lilac Cap Vial with Insulin, 0.50 ml
1 × Natural Cap Vial with Hydrocortisone, 0.50 ml
1 × Red Cap Vial with GA-1000, 0.50 ml