FBM® Fibroblast Growth Basal Medium
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3131
CA10065-254EA 262.68 CAD
CA10065-254
FBM® Fibroblast Growth Basal Medium
Cell Culture Media
FBM® basal medium is a component sold separately. This is part of the FGM® -2 and FGM® -3 Fibroblast Growth Media BulletKits® (Catalog Numbers CC-3132 and CC-4526, respectively) which consist of basal medium and supplements packaged together.
FGM®-2 Fibroblast Growth Medium-2 (CC-3132) was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. FGM®-2 Medium contains 2% serum.
FGM®-3 Fibroblast Growth Medium-3 (CC-4526) was developed specifically for cardiac fibroblasts. FGM®-3 Medium contains contains 10% serum.
Delivery information: Content: 1 × FBM® Basal Medium, 500 ml.