BulletKit® EGM® Endothelial Cell Growth Medium
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-3124
CA94045-820EA 436.48 CAD
CA94045-820
BulletKit® EGM® Endothelial Cell Growth Medium
Cell Culture Media
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types.
Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics® Primary Cells.
The following Endothelial Cell Growth Medium BulletKits® are available:
EGM®-2 BulletKit® (CC-3162) – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
EGM® BulletKit® (CC-3124) – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
EGM®-Plus BulletKit® (CC-5035) – Enhanced version of EGM® Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM® Medium.
EGM®-2MV BulletKit® (CC-3202) – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
Lonza guarantees the performance of Clonetics®/Poietics® Cells only if appropriate Clonetics®/Poietics® Media and Reagents are used exclusively and the recommended storage and use protocols are followed.
Delivery information: Content:
1 × EBM® Basal Medium (CC-3121), 500 ml
1 × EGM® SingleQuots® Supplement Pack (CC-4133) containing:
1 × Green Cap Vial with hEGF 0.50 ml
1 × Natural Cap Vial with Hydrocortisone 0.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml
1 × Bottle BBE 2.00 ml
1 × Blue Cap Vial with Ascorbic Acid 0.50 ml
1 × Bottle FBS 10.00 ml