HRE – Human Renal Epithelial Cells
Supplier: Lonza
CC-2556
CA10064-250EA 1874.28 CAD
CA10064-250
HRE – Human Renal Epithelial Cells
Cells
Lonza's Human Renal Cells include cells isolated from various normal human kidney tissue specific to each cell type. Human Renal Proximal Tubule Cells are isolated from the proximal tubule and are ideal for drug transporter studies.
Human Renal Epithelial Cells (HRE) are comprised of cells from the cortex and glomerular region. Human Renal Cortical Cells (HRCE) are from proximal and distal tubules.
Select lots of renal proximal tubule epithelial cells isolated in the absence of Gentamycin-Amphotericin are available. These lots are isolated in Doxycycline. Contact your local account representative to purchase these lots.
Renal epithelial cells display a polarized morphology, which is essential for their function. They reabsorb nearly all of the glucose and amino acids in the glomerular filtrate, while allowing other substances of no nutritional value to be excreted in the urine. Physiological processes like osmoregulation and excretion can be studied with HRE in vitro. Human renal epithelial cells are also capable of internalizing bacteria and are located near injury sites in a variety of congenital, metabolic and inflammatory diseases.
Delivery information: Content: 1 × Cryopreserved Ampule with ≥500000 cells/vial.