NHBE – Human Bronchial/Tracheal Epithelial Cells without Retinoic Acid
Supplier: Lonza
CC-2541
CA10065-216EA 2095.48 CAD
CA10065-216
NHBE – Human Bronchial/Tracheal Epithelial Cells without Retinoic Acid
Cells
Normal and Diseased Bronchial Epithelial (NHBE and DHBE) cells are isolated from epithelial lining of airways above bifurcation of the lungs. Lonza's Normal Human Bronchial Epithelial Cells(NHBE) without retinoic acid are available. Certain lots with Small Airway epithelial cells(CC-2547) or normal human lung fibroblasts(CC-2512) from matched donors are available.
  • All cells test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Cryopreserved human bronchial epithelial cells without retinoic acid are guaranteed through 15 population doublings.
Select Applications with Lonza's Human Bronchial Epithelial Cells :
A Rotating Bioreactor for Scalable Culture and Differentiation of Respiratory Epithelium. Respiratory epithelium is difficult to grow as it requires a well-maintained polarizing air‐liquid interface (ALI) to maintain differentiation. Raredon et al. describe a scalable and easy method to conduct air-liquid interface with Lonza's bronchial epithelial cells guaranteed for ALI differentiation (Cat. No. CC-2540S).
Cell concentration and space for growth are required for productive NHBE cell branching in vitro. Click to read this article by Hagiwara et.al that shows how co-cultures with HUVECs in 3D culture influence reconstruction of branched tubular structures in lung epithelial cells.
Wu, et al. reports for the first time that HBE cells when grown in B-ALI®Medium can differentiate into 3D glandular acinar structures, likesalivary and mammary epithelial. The cells were grown on the basement membrane matrix Matrigel under select conditions and have shown to exhibit formation of brochospheres or tracheospheres with ciliated lining.
Ordering information: Content: 1 × Cryopreserved ampule containing ≥500000 cells. Certificate of Analysis provided for every cell lot.