KGM® Gold Keratinocyte Growth Medium BulletKit®, Calcium Free
Supplier: LONZA PHARMA - BIOSCIENCE CA
00195769
CA10063-844EA 294.28 CAD
CA10063-844
KGM® Gold Keratinocyte Growth Medium BulletKit®, Calcium Free
Cell Culture Media
A new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
KGM-Gold® Calcium free keratinocyte growth medium is designed to support both clonal growth and high density keratinocyte proliferation. The medium supports superior growth and morphology of keratinocytes.
Furthermore, quality control at Lonza requires that keratinocytes pass strict morphological criteria specifying the minimal presence of enlarged and vacuolated cells, enabling Lonza keratinocytes to be used by the researcher for ≥15 population doublings.
Delivery information: Content:
1×Bottle Basal medium 500 ml.
1×Natural cap vial with hydrocortisone 0.50 ml.
1×Natural cap vial with transferrin 0.50 ml.
1×Amber vial with epinephrine 0.25 ml.
1×Red cap vial with GA-1000 0.50 ml.
1×Orange cap vial with BPE 2.00 ml.
1×Green cap vial with hEGF 0.50 ml.
1×Lilac cap vial with insulin 0.50 ml.