D-HLF-COPD – Diseased Human Lung Fibroblast Cell – COPD
Supplier: LONZA PHARMA - BIOSCIENCE CA
00195277
CA10063-836EA 2150.78 CAD
CA10063-836
D-HLF-COPD – Diseased Human Lung Fibroblast Cell – COPD
Cells
Diseased fibroblasts are isolated from adult COPD donors. The most abundant cell type in lung interstitium is fibroblasts.
They resemble ordinary fibroblasts but have some distinguishing features, for example, they have long branching processes and gap junctions. Their principle function is production of type III collagen, elastin, and proteoglycans of the extracellular matrix of the alveolar septa.
Pulmonary fibroblasts (PF) play an important role in the repair and remodeling processes following injury. The controlled accumulation of fibroblasts to sites of inflammation is crucial to effective tissue repair after injury. Either inadequate or excessive accumulation of fibroblasts could result in abnormal tissue function. For example, the excess proliferation of fibroblasts contributes to the adventitial thickening observed during the development of hypoxia-induced pulmonary hypertension.
Cryopreserved diseased lung fibroblast cells stain negative for von Willebrand factor VIII, cytokeratins 18 and 19, and alpha smooth muscle actin. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery information: Content: 1×cryopreserved ampule containing ≥500000 cells. A Certificate of Analysis is provided for each cell lot purchased.