Nucleofector® Mouse Embryonic Fibroblast Kit 2
Supplier: LONZA PHARMA - BIOSCIENCE CA
VPD-1005
CA10064-436EA 1269.93 CAD
CA10064-436
Nucleofector® Mouse Embryonic Fibroblast Kit 2
Transfection Reagents
The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell.
  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
The mouse embryonic fibroblast Nucleofector® kits are suitable for efficient transfection of primary mouse embryonic fibroblasts (MEF). Due to significant diversity between different MEF clones, we have developed two Nucleofector® Kits (1 and 2) and an easy-to-use Nucleofector® starter kit to determine which of the two kits is optimal for a specific MEF clone.
Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
Delivery information: Content: 2.25 ml Nucleofector® solution for mouse embryonic fibroblast cells, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.