Nucleofector® Human Natural Killer Cell Kit
Supplier: Lonza
Nucleofector®
VPA-1005
CA10064-616EA 1269.93 CAD
CA10064-616
Nucleofector® Human Natural Killer Cell Kit
Transfection Reagents
The Nucleofector® 2b system (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary natural killer (NK) cells.
  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
Delivery information: Content: 2.25 ml Nucleofector® solution for human natural killer cells, 0.5 ml supplement 1, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.