BdSMC – Human Bladder Smooth Muscle Cells
Supplier: Lonza
CC-2533
CA10064-212EA 2196.2 CAD
CA10064-212
BdSMC – Human Bladder Smooth Muscle Cells
Cells
Clonetics® Human Bladder Smooth Muscle Cells (BdSMC) are isolated from specific tissues surrounding the bladder. Cells are shipped in passage 3.
BdSMC are guaranteed through 10 population doublings and to stain positive for alpha smooth muscle actin and negative for von Willebrand Factor VIII. Cells test negative for mycoplasma, bacteria, yeast and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Applications: Overactive bladder causes, cancer, urologic disease, reproductive disease.
Delivery information: Content: 1×cryopreserved Ampule with ≥500000 cells/vial.