Power Rails for VWR® Lab Benches
Supplier: Treston
14-91118931 14-91118933 14-91118935 14-91118937 14-91118939 14-91118961 14-91118963 14-91118965 14-91118967 14-91118969
77394-590EA 367.98 CAD
77394-590 77394-592 77394-594 77394-596 77394-598 77394-600 77394-602 77394-604 77394-606 77394-608
Power Rails for VWR® Lab Benches
Power Supplies
Power rails for attachment to slotted uprights on VWR® Lab Benches. Slim black power rail allows for easy access to outlets while working.
  • Manufactured in the USA
  • Available in M30, M36, M48, M60, or M72 to expand the length of slotted upright bays
  • Built-in mounting brackets; no assembly required
  • Available in 15, 20 A and twist lock plug (upon request)
  • Surge and non-surge available (upon request)
  • Black power rails with other colors available as needed
  • Additional power/data configurations are possible
  • One year warranty
For use with any VWR® C-Leg or 4-More Product Information