Sterilization Pouches, Self Seal Paper
Supplier: O&M HALYARD CANADA, INC CA
48538 48539 48540 48541 48542 48543 48544 48545 48546 48547 48548 48549 48550 48559 48560 48561
77322-692CS 284.02 CAD
77322-692 77322-694 77322-696 77322-698 77322-700 77322-702 77322-704 77322-706 77322-708 77322-710 77322-712 77322-714 77322-716 77322-734 77322-736 77322-738
Sterilization Pouches, Self Seal Paper
Bags Sterilization Pouches
HALYARD* Paper-Film single-use sterilization pouches are intended to be used to enclose another medical device, in a single or double pouch configuration, that is to be sterilized by a health care professional via steam and EO.
  • Single-use disposable
  • Premium 68 GSM paper consisting entirely of primary virgin fiber
  • Excellent bacterial barrier properties that allow proper sterilant penetration
  • Provide exceptional clean peeling performance and high seal strength
  • Multilayer transparent film provides puncture and abrasion resistance plus a high-quality seal
More Product Information