X-TRA TRACTION* Shoe Covers
Supplier: O&M HALYARD CANADA, INC CA
69252 69254
77322-740CS 133.16 CAD
77322-740 77322-742
X-TRA TRACTION* Shoe Covers
Disposable Garments
The Halyard health footwear product line features innovative fabrics and styling to deliver the right amount of protection for the task.
  • Sewn seams
  • Available in universal and extra large sizes
  • Breathable, fluid-repellent 3-layer SMS fabric
  • Adhesive traction strips
X-TRA TRACTION* shoe covers are designed to protect against light fluid contact.