Sterilization Pouches, Heat Seal Paper
Supplier: O&M HALYARD CANADA, INC CA
48492 48493 48494 48495 48496 48507 48508 48509 48510 48511 48512 48513 48514 48515 48516 48555 48556 48497 48498 48499 48500 48501 48502 48503 48504 48505 48506 48517 48518 48519 48520 48521 48522 48523 48524 48525 48526 48557 48558
77322-600CS 356.16 CAD
77322-600 77322-602 77322-604 77322-606 77322-608 77322-630 77322-632 77322-634 77322-636 77322-638 77322-640 77322-642 77322-644 77322-646 77322-648 77322-726 77322-728 77322-610 77322-612 77322-614 77322-616 77322-618 77322-620 77322-622 77322-624 77322-626 77322-628 77322-650 77322-652 77322-654 77322-656 77322-658 77322-660 77322-662 77322-664 77322-666 77322-668 77322-730 77322-732
Sterilization Pouches, Heat Seal Paper
Bags Sterilization Pouches
HALYARD* paper-film single-use sterilization pouches are intended to be used to enclose another medical device, in a single or double pouch configuration, that is to be sterilized by a health care professional via steam and EO.
  • Single-use disposable
  • Premium 68 GSM paper consisting entirely of primary virgin fiber
  • Excellent bacterial barrier properties that allow proper sterilant penetration
  • Provide exceptional clean peeling performance and high seal strength
  • Multilayer transparent film provides puncture and abrasion resistance plus a high-quality seal
More Product Information