α-Pinene, natural isolate

Supplier: Ricca Chemical
Danger

RCCT9040-20C6 RCCT9040-750C6 RCCT9040-100C6
RCCT9040-20C6EA 34.76 CAD
RCCT9040-20C6 RCCT9040-750C6 RCCT9040-100C6
α-Pinene, natural isolate
α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001339
CAS Number: 80-56-8
UN: 2368
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR