α-Terpinene, natural isolate

Supplier: Ricca Chemical
Danger

Synonyms: 1-Isopropyl-4-methylcyclohexa-1,3-dienep-Mentha-1,3-diene

RCCT9032-750C6 RCCT9032-100C6 RCCT9032-20C6
RCCT9032-20C6EA 62 CAD
RCCT9032-20C6 RCCT9032-750C6 RCCT9032-100C6
α-Terpinene, natural isolate
α-Terpinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001534
CAS Number: 99-86-5
UN: 2319
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR