(±)-Camphene, natural isolate

Warning

Synonyms: Camphene, (±)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, (±)-3,3-Dimethyl-2-methylennorcamphane

RCCT9028-750C6 RCCT9028-100C6 RCCT9028-20C6
RCCT9028-750C6EA 1452.02 CAD
RCCT9028-750C6 RCCT9028-100C6 RCCT9028-20C6
(±)-Camphene, natural isolate
(±)-Camphene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 318
Flash Pt: 34
MDL Number: MFCD00066603
CAS Number: 79-92-5
Merck Index: 12,01777

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR