Geraniol, natural isolate

Supplier: Ricca Chemical
Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

RCCT9012-100C6 RCCT9012-20C6 RCCT9012-750C6
RCCT9012-100C6EA 225.94 CAD
RCCT9012-100C6 RCCT9012-20C6 RCCT9012-750C6
Geraniol, natural isolate
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR