pH Electrodes, LE427 / LE438, Mettler-Toledo
Supplier: Mettler Toledo
30247153 30259840
CA10821-384EA 449.43 CAD
CA10821-384 CA10821-386
pH Electrodes, LE427 / LE438, Mettler-Toledo
Electrodes pH Electrodes
Waterproof pH electrodes for use with portable FiveGo™ meters.
LE438: 3-in-1 plastic pH electrode with integrated temperature sensor for pH field use, samples with fluctuating temperatures; low-maintenance.
LE427: The spear tip design makes pH electrode ideal for solid samples such as meat, fruit, cheese, soil and many more; low-maintenance.