HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.5% (by titrimetric analysis), cGMP-manufactured

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

H1804-1KGBL H1804-12KGBL H1804-25KGBL
76472-812EA 2104.09 CAD
76472-812 76472-814 76472-816
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.5% (by titrimetric analysis), cGMP-manufactured
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

HEPES, cGMP Manufactured is a zwitterionic biological buffer and is one of Good's buffers widely used in cell culture. Non-graded HEPES, cGMP Manufactured supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₈N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9

Order Now

Specification Test Results

Assay (titration) min. 99.5 %
pH OF A 5% AQUEOUS SOLUTION 4,5-6,5
pKa @ 25 C 7,35-7,75
Absorbance @ 260 nm (0.05 M in water) 0,05 AU
Absorbance @ 280 nm (0.05 M in water) 0,03 AU
WATER (KF) max. 0,5%
HEAVY METALS (as Pb) max. 5ppm
INSOLUBLE MATTER max. 0,1 %
TOTAL AEROBIC MICROBIAL COUNT max. 100cfu/g
TOTAL YEAST AND MOLD COUNT max. 100cfu/g
BACTERIAL ENDOTOXIN as reported


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR