Brady® FR22 Fixed RFID Reader Accessories
Supplier: Brady Worldwide
311922 313687 320033 320034 311927 311928 311929 311930 311931 311940 311941 311943
77322-116EA 311.28 CAD
77322-116 77322-140 77322-144 77322-146 77322-118 77322-120 77322-122 77322-124 77322-126 77322-130 77322-132 77322-134
Brady® FR22 Fixed RFID Reader Accessories
Barcode and RFID Systems
Antennae, cables, power supplies, and other accessories are available.
MUX 16-Port Multiplexer Antenna Enabler for FR22 Fixed RFID Reader - Extend the number of available RFID readings zones with the 16 Port Multiplexer. Use with the FR22 Fixed RFID Reader to extend the number of antenna ports from 4 to 16. Integrates fully with the FR22 fixed RFID reader. External antenna: 16 ports RP-SMA. Frequency: Wide band 865 MHz to 928 MHz.
GA30 RFID Antenna for FR22 Fixed RFID Reader - A high-gain, general purpose antenna, the GA30 RFID Antenna in perfect for applications requiring long read ranges.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable