Luer Lock Syringes
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
SLL101103 SLL101110 SLL101101 SLL101105 SLL101120
76837-026BX 35.22 CAD
76837-026 76837-028 76837-030 76837-025 76837-029 76837-033
Luer Lock Syringes
Syringes General-Purpose Syringes
General-purpose syringes available in 1, 3, 5, 10 and 20 ml barrel volumes for clinical and veterinary use.
  • EtO sterilized
  • Pyrogen-free
  • Highly transparent barrel for maximum visibility
  • Excellent lubrication performance
  • Plunger retention mechanism prevents accidental plunger withdrawal
These syringes are non-reactive and safe for use in medical procedures. The barrel and plunger are made from polypropylene, and the piston is made from polyisoprene. All syringes do not contain any DEHP, BPA, or styrene. This makes them suitable for clinical use including injecting fluid into the body and collecting fluid from the body.
Certifications: Meets ISO standards, FDA approved.
Packaging: Syringes are sterilized, individually wrapped.