Allegra V-15R Benchtop Centrifuges 200/220-240 V, Beckman Coulter
Supplier: Beckman Coulter
C69864 C69865 C69878 C69879 C63377 C63383 C63389 C63423 C63429 C63435 C63441 C63447
76628-200EA 31446.99 CAD
76628-200 76628-202 76628-208 76628-210 76628-184 76628-186 76628-188 76628-190 76628-192 76628-194 76628-196 76628-198
Allegra V-15R Benchtop Centrifuges 200/220-240 V, Beckman Coulter
Centrifuges Benchtop Centrifuges
The Allegra V-15R benchtop centrifuges bring more versatility to the laboratory bench. These compact, refrigerated centrifuges are ideal for evolving laboratories.
  • 200/220-240 V models
  • Free up precious benchtop space
  • Support multiple workflows with just one instrument
  • Advanced temperature management
  • Wide choice of centrifuge packages available to suit various applications
The Allegra V-15R centrifuges maintain a consistent temperature, allowing you to spin faster and longer without overheating your samples. All while Rapid Temp and Eco Mode allow forMore Product Information