General Purpose Water Baths, Boekel Scientific
Supplier: Boekel
290300-2 290100-2 290200-2
97004-334EA 4331.92 CAD
97004-334 97004-336 97004-340
General Purpose Water Baths, Boekel Scientific
Baths Water Baths, Unstirred
The Boekel Scientific family of Water Baths offers a variety of sizes to meet all of your needs and is ideal for general laboratory applications
  • 230 V models, not for use in USA
Equipped with an adjustable over temperature safety cutout which sounds an audible alarm and shuts the heater off helps protect your samples while the recessed control panel helps protect against spills. Also featured is a seamless passivated tank to prevent against rust and corrosion.
The space saving design allows for the most efficient use of your bench space and the Microprocessor Controller makes for easy temperature selection, rapid heat-up and excellent stability. The special boiling mode is a convenient feature for melting agar as well as other applications that require a boiling temperature (to utilize the Boiling Mode, you will need to purchase the Stainless Steel Gabled Lid accessory).
Certifications: UL/ CSA Listed, CE Certified.
Ordering information: Unit includes a Polycarbonate Lid, Stainless Steed Diffuser Tray, Instruction Manual and Power Cord. Stainless Steel Gabled Lid sold separately. To utilize the Boiling Mode, you will need to purchase the Stainless Steel Gabled Lid accessory. All units include a two-year warranty on parts and labor.
Caution: 230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (97004-338, 97004-342 and 97004-344) are also available, please see separate product page.