biOrb Air 30
Supplier: GEOGLOBAL PARTNERS
71748 71749 72558 77668 77669 85240
470352-580EA 509.4 CAD
470352-580 470352-584 470352-586 470352-590 470352-592 470352-588
biOrb Air 30
The ultimate terrarium for plant enthusiasts.
  • Perfect lighting
  • Maintains humidity levels
  • Unique design and construction
  • Two year warranty
Showcase and grow healthy tropical, temperate, or semi-arid plants in this fully automated terrarium. The newest model, the AIR 30, features a compact size, making it ideal for desktop or tabletop display.
This terrarium senses when the humidity drops and gently mists and circulates air to stay within the selected relative humidity (RH) range. Use the included remote control to choose between three settings: low for semi-arid plants, moderate for temperate plants with no specific needs, and high for tropical plants. Natural daylight is not necessary as the lighting is arranged in an LED array at the top of the sphere. This emits the perfect light output that promotes natural growth and prevents plants from bending towards a single light source. Use the remote to control the brightness or pick an 8-hour, 10-hour, 12-hour, or 14-hour light cycle, with each cycle simulating sunrise, daylight, dusk, and nighttime.
Enjoy a 360 degree view of this mini paradise through its globe-like shape. Its acrylic material is 10 times stronger than glass, 50% lighter, and has a 23% transparency rating for a clear view. Plus, the low heat loss of acrylic makes it perfect for regulating the internal climate of the terrarium.