Staticide® 8124 Disposable Heel Strap
Supplier: ACL Staticide
Staticide®
8124
77200-602PK 64.39 CAD
77200-602
Staticide® 8124 Disposable Heel Strap
Grounders Heel and Toe Grounders
The easy to use ACL staticide disposable heel strap is an economical solution for plant visitors and other temporary workers who enter electrostatic protected areas.
  • 99% polyester, 1% conductive yarn
  • Resistance: 10⁵ ohms
These disposable heel straps are packaged 100 strips per bag.