Staticide® Dualmat™ ESD Safe Tray Liners, Gray
Supplier: ACL Staticide
7375DGYM1813RE 7375DGYM2416RE
77200-660EA 56.66 CAD
77200-660 77200-662
Staticide® Dualmat™ ESD Safe Tray Liners, Gray
Surface Protectors Bench Protectors
The Dualmat™ 06 Tray liners are durable, static dissipative, and offer reliable static control.
  • Two-layer, conductive/dissipative design
  • Includes 4 oz. mat cleaner
  • Embossed surface
  • Made in USA
Mats consist of two layers, with a conductive bottome and dissipative top side. Bottom side resistance measures less than 5×10⁵ ohms. Top side resistance measures 10⁶ to 10⁸ ohms.
Certifications: Meets ANSI/ESDA S20.20.