HydroSpec® Water Standards for Karl Fischer Titration
Supplier: Ricca Chemical
RK410000-10X5R
RK420000-10X5R
RK430000-10X5R
RCRK410000105REA
371.3
CAD
RCRK410000105R
RCRK420000105R
RCRK430000105R
HydroSpec® Water Standards for Karl Fischer Titration
Standards
Water Determination (by Karl Fischer method) Standards
Achieve high performance and reliable results. Hydrospec® supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.
- Reliable, Safe, Pyridine free
- Stable Formulations and Comprehensive Chemical Compatability
- 5 year shelf life
- Technical Support Every Step of the Way
- Tight specifications for More Accurate Reults and Low Lot-to-Lot Variability