Eisco Glass Gilson Separatory Funnels with Glass Stopcock
Supplier: Wards
CH0487E CH0487C CH0487A CH0487B CH0487D
470343-972EA 122.6 CAD
470343-972 470344-662 470345-188 470345-250 470346-028
Eisco Glass Gilson Separatory Funnels with Glass Stopcock
Funnels Separating Funnels
Ungraduated Gilson separatory funnels with ground cones, plastic stoppers, and glass key stopcocks, made of borosilicate 3.3 glass.
  • Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Gilson style features a long delivery stem for good control of the liquid/liquid interface
  • Fitted with a glass key stopcock for precise drop-wise dispensing of the denser separated liquid
  • With ground glass cone with and interchangeable plastic stopper
  • Ungraduated
Gilson style Separating Funnels have a long delivery stem for good control of the liquid/liquid interface. They feature a ground cone, plastic stopper, and glass key stopcock. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C