Eisco Glass Separatory Funnels with Glass Stopcock
Supplier: Wards
CH0482E CH0482D CH0482F CH0482C CH0482A CH0482B
470343-684EA 72.35 CAD
470343-684 470343-932 470344-034 470345-234 470345-414 470345-832
Eisco Glass Separatory Funnels with Glass Stopcock
Funnels Separating Funnels
Separatory funnels with ground cones, plastic stoppers, and glass key stopcocks, made of borosilicate 3.3 glass.
  • Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Fitted with a glass key stopcock for precise drop-wise dispensing of the denser separated liquid
  • Features a ground glass cone with and interchangeable plastic stopper
  • Ungraduated
Ungraduated, pear-shaped separating funnels are made of high quality borosilicate glass. They feature a ground cone, plastic stopper, and glass key stopcock. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C