Eisco Plastic Separatory Funnels
Supplier: Wards
CH0480A CH0480C CH0480B
470343-560EA 26.4 CAD
470343-560 470343-704 470344-278
Eisco Plastic Separatory Funnels
Funnels Separating Funnels
Polypropylene separatory funnels have a screw cap and key stopcock fitted with a self-lubricating PTFE plug.
  • Made of premium-quality polypropylene plastic, with superior optical clarity, chemical durability, and heat tolerance
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Polypropylene key stopcock fitted with a self-lubricating PTFE plug push-fitted to the stem of the funnel for easy and precise flow control during separation procedures
  • Leak-proof threaded screw cap for protecting contents
  • Ungraduated