Borosil® Measuring Cylinders, Graduated, Pour-Out Single TC, Class B
Supplier: Foxx Life Sciences
3027005 3027006 3027009 3027012 3027016 3027021 3027024 3027029 3027030
76383-216CS 176.6 CAD
76383-216 76383-218 76383-220 76383-222 76383-224 76383-226 76383-228 76383-230 76383-232
Borosil® Measuring Cylinders, Graduated, Pour-Out Single TC, Class B
Cylinders Measuring Cylinders
Borosil® graduated measuring cylinder, pour out, single metric, Class B accuracy as per ASTM E 1272 with hexagonal base.
  • Calabrated per Class B tolerences
  • Graduation in amber enamel
  • 3.3 borosilicate glass
  • Sturdy hexagonal base
Borosil® measuring cylinders comply with ASTM E 1272. Calibrated as per Class B tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with amber enamel and the cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability.
Certifications: In compliance with USP tolerance standards. ASTM 1272.