Kimtech™ Polaris™ Nitrile Examination Gloves, 9.5", Ambidextrous
Supplier: Microflex
62770 62771 62772 62773 62774
77308-536BX 42.81 CAD
77308-536 77308-538 77308-540 77308-542 77308-544
Kimtech™ Polaris™ Nitrile Examination Gloves, 9.5", Ambidextrous
Gloves
Kimtech™ is s partner in driving progress. As part of Kimberly Clark's 150-year legacy of innovation and partnership with the scientific community, we're proud to present the next step into the future; Kimtech™ Polaris™ nitrile gloves.
  • Polaris™ nitrile examination gloves are tested for use with Fentanyl and 24 chemotherapy drugs
  • Gloves meet USP 800 guidelines when used for non-sterile compounding applications
  • Certified by US Ergonomics
  • Ambidextrous with textured fingertips to enhance dry and wet grip
  • Gloves meet FDA food contact requirements in 21 CFR 177.2600
  • Strong and durable with superior quality at AQL 0.65 (exceeds the ASTM AQL 2.5 standard for critical defects or pinholes)
Tested against some of the harshest chemicals used in the work environment, Kimtech™ Polaris™ nitrile examination gloves meet and exceed the industry standards for examination gloves. The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ examination glove portfolio, and has been tested for use with chemotherapy drugs and fentanyl. With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
Eligible for recycling through The RightCycle™ Program by Kimberly-Clark Professional™, 100% of the non-hazardous Kimtech™ Polaris™ nitrile examination gloves can be diverted from the landfill. Offering reliable protection, durability, and comfort, these gloves are built for science, made for you.
Packaging: Glove sizes XS to L are packaged 100 per box, 10 boxes per case. The XL size gloves are packaged 90 per box, 10 boxes per case.