Kimtech™ Polaris™ Xtra Nitrile Examination Gloves, 12", Ambidextrous
Supplier: Microflex
62760 62761 62762 62763 62764
77308-526BX 38.51 CAD
77308-526 77308-528 77308-530 77308-532 77308-534
Kimtech™ Polaris™ Xtra Nitrile Examination Gloves, 12", Ambidextrous
Gloves
Kimtech™ is a partner in driving progress. As part of Kimberly Clark's 150-year legacy of innovation and partnership with the scientific community, we're proud to present the next step into the future; Kimtech™ Polaris™ Xtra nitrile gloves.
  • Polaris™ Xtra nitrile examination gloves are tested for use with Fentanyl and 24 chemotherapy drugs
  • Gloves meet USP 800 guidelines when used for non sterile compounding applications
  • Certified by US Ergonomics
  • Ambidextrous with textured fingertips to enhance dry and wet grip
  • Gloves meet FDA food contact requirements in 21
More Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Finish Textured fingertips
Cuff style Beaded
Cuff thickness 4.33 mil
Finger thickness 7.48 mil
Food contact approved +