Eisco Glass Filter Funnels - Heavy

Supplier: Wards
CH0458C CH0458D
470344-588EA 9.7 CAD
470344-588 470345-810
Eisco Glass Filter Funnels - Heavy
Funnels Filtration Funnels Filter Funnels
Heavy, thick-walled filter funnels with 60° angles.

  • Made of high quality Borosilicate 3.3 glass that is built to last and withstand multiple uses
  • Heavy construction provides greater mechanical strength and prolonged life with harder than normal usage
  • Features thick, uniform walls that allow for increased durability, ease of use and continual flow of liquid

Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.

Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR